Human Recombinant CD40 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; FACS analysis shows that Biotinylated Human CD40, Fc,Avitag (Cat. No. TN5-H82F9) can bind to 293T cells overexpressing human CD40L. The concentration of Biotinylated Human CD40 is 0.1 μg/mL (Routinely tested).
- Gene ID:P25942-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:p50|CD40|MGC9013|Bp50|TNFRSF5|CDW40
- Protein/Peptide Name:CD40
- Purity:>95%
- Molecular Weight:48.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together