Human Recombinant CD30 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.01 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CD30, Llama IgG2b Fc Tag, low endotoxin (Cat. No. TN8-H5250) at 0.5 μg/mL (100 μL/well) can bind Anti-CD30 Mab, Human IgG1 with a linear range of 0.1-1.6 ng/mL (QC tested).; Loaded Human CD30 Ligand, His Tag, low endotoxin (Cat. No. CDL-H524b) on HIS1K Biosensor, can bind Human CD30, Llama IgG2b Fc Tag, low endotoxin (Cat. No. TN8-H5250) with an affinity constant of 55.5 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P28908-1(C297R)
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:Ki-1|CD30|D1S166E|TNFRSF8
- Protein/Peptide Name:CD30
- Purity:>95%
- Molecular Weight:66.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together