Human Recombinant CD19 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized FMC63 at 1 μg/mL (100 μL/well) can bind Human CD19 (20-291), Fc Tag, low endotoxin (Super affinity) (Cat. No. CD9-H5251) with a linear range of 1-31 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human CD19 (20-291), Fc Tag, low endotoxin (Super affinity) (Cat. No. CD9-H5251) at 5 μg/mL (100 μL/well) can bind FMC63 (Mouse IgG2a) with a linear range of 0.6-5 ng/mL (Routinely tested).; Human CD19 (20-291), Fc Tag, low endotoxin (Super affinity) (Cat. No. CD9-H5251) captured on CM5 chip via Anti-Human lgG Fc antibodies surface, can bind FMC63 MAb (mouse lgG2a) with an affinity constant of 0.17 nM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Human CD19 (20-291), Fc Tag, low endotoxin (Super affinity) (Cat. No. CD9-H5251) on AHC Biosensor, can bind FMC63 MAb (mouse lgG2a) with an affinity constant of 0.483 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; 293 cells were transfected with FMC63-scFv and RFP tag. 2e5 of the cells were first stained with B. Human CD19 (20-291) Protein, Fc Tag, low endotoxin (Super affinity) (Cat. No. CD9-H5251, 3 µg/ml) and C. Human Fc Tag Protein Control, followed by FITC-conjugated Anti-human IgG Fc Antibody. A. Non-transfected 293 cells and C. Human Fc Tag Protein Control were used as negative control. RFP was used to evaluate CAR (anti-CD19-scFv) expression and FITC was used to evaluate the binding activity of Human CD19 (20-291) Protein, Fc Tag, low endotoxin (Super affinity) (Cat. No. CD9-H5251).
- Gene ID:P15391-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CVID3|MGC12802|B4|CD19
- Protein/Peptide Name:CD19
- Purity:>90%
- Molecular Weight:56.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together