Human Recombinant BTN3A1 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Gene ID:O00481-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:BTF5|CD277|BTN3A1
- Protein/Peptide Name:BTN3A1
- Purity:>95%
- Molecular Weight:52.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together