Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant BTLA (from HEK293)
Human Recombinant BTLA (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Human Recombinant BTLA (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human HVEM, Fc Tag (Cat. No. HVM-H5258) at 10 μg/mL (100 μL/well) can bind Biotinylated Human BTLA (31-134), Fc,Avitag (Cat. No. BTA-H82F8) with a linear range of 0.3-5 μg/mL (QC tested).
  • Gene ID:
    Q7Z6A9
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    BTLA|CD272
  • Protein/Peptide Name:
    BTLA
  • Purity:
    >95%
  • Molecular Weight:
    40.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together