Human Recombinant B7-H6 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human NKp30, Fc Tag (Cat. No. NC3-H5259) at 10 μg/mL (100 μL/well) can bind Human B7-H6, His Tag (Cat. No. B76-H52H8) with a linear range of 0.016-0.25 μg/mL (QC tested).
- Gene ID:Q68D85-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:B7-H6|B7 Homolog 6|NCR3LG1
- Protein/Peptide Name:B7-H6
- Purity:>90%
- Molecular Weight:28.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together