Human Recombinant B7-H3 (4Ig) (from HEK293)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) at 2 μg/mL (100 μL/well) can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) with a linear range of 1-10 ng/mL (Routinely tested).; Human B7-H3 (4Ig), His Tag (Cat. No. B7B-H52E7) inhibits Anti-CD3-induced proliferation of PBMC. The EC50 for this effect is 7.95 μg/mL (Routinely tested).
- Gene ID:Q5ZPR3-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:B7 homolog 3|B7-H3|UNQ309/PRO352|4Ig-B7-H3|PSEC0249|CD276
- Protein/Peptide Name:B7-H3 (4Ig)
- Purity:>95%
- Molecular Weight:48.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT