Human Recombinant ANGPTL7 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human LILRB2, Fc Tag (Cat. No. CDD-H5259) at 5 μg/mL (100 μL/well) can bind Biotinylated Human ANGPTL7, Mouse IgG2a Fc,Avitag (Cat. No. AN7-H82F3) with a linear range of 0.156-2.5 μg/mL (QC tested).
- Gene ID:O43827-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CDT6|ANGPTL7
- Protein/Peptide Name:ANGPTL7
- Purity:>90%
- Molecular Weight:65.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together