Cynomolgus Recombinant TIM-3 (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus TIM-3, Fc Tag (Cat. No. TI3-C525b) at 2μg/mL (100 μL/well) can bind Anti-TIM3 Mab, Human IgG4 with a linear range of 0.5-4 ng/mL (QC tested).
- Gene ID:G7P6Q7
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:TIMD3|FLJ14428|KIM3|TIM3|HAVCR2
- Protein/Peptide Name:TIM-3
- Purity:>85%
- Molecular Weight:45.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together