Cynomolgus Recombinant TIGIT (from HEK293)
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus / Rhesus macaque TIGIT, His Tag (HPLC-verified) (Cat. No. TIT-C5223) at 5 μg/mL (100 μL/well) can bind Human CD155, Fc Tag (Cat. No. CD5-H5251) with a linear range of 0.078-2.5 μg/mL (QC tested).; Loaded Human CD155, Fc Tag (Cat. No. CD5-H5251) on Protein A Biosensor, can bind Cynomolgus / Rhesus macaque TIGIT, His Tag (HPLC-verified) (Cat. No. TIT-C5223) with an affinity constant of 0.61 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:G7NXM4-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:VSTM3|TIGIT|VSIG9
- Protein/Peptide Name:TIGIT
- Purity:>90%
- Molecular Weight:15.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
Specifications
Frequently Bought Together