Cynomolgus Recombinant LAG-3 (from HEK293)
103738-204
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:100 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Cynomolgus LAG-3, Mouse IgG2a Fc tag (Cat. No. LA3-C52A0) at 2 μg/mL (100 μL/well) can bind Madarex LAG-3 Mab, Human IgG1 with a linear range of 20-78 ng/mL (QC tested).
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:FDC|CD223|LAG3
- Protein/Peptide Name:LAG-3
- Purity:>90%
- Molecular Weight:73.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103738-204