Order Entry
Puerto Rico
ContactUsLinkComponent
Cynomolgus Recombinant Fc gamma RIII / CD16 (from HEK293)
Cynomolgus Recombinant Fc gamma RIII / CD16 (from HEK293)
Certificates
Cynomolgus Recombinant Fc gamma RIII / CD16 (from HEK293)

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Rituximab at 5 μg/mL (100 μL/well) can bind Cynomolgus CD16, His Tag (BLI verified) (Cat. No. FC6-C52H9) with a linear range of 39-313 ng/mL (QC tested).
    ; Captured Cynomolgus CD16, His Tag (BLI verified) (Cat. No. FC6-C52H9) on CM5 chip via anti-His antibody can bind MabThera® (Rituximab) with an affinity constant of 0.206 μM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Cynomolgus CD16, His Tag (BLI verified) (Cat. No. FC6-C52H9) on HIS1K Biosensor, can bind Rituximab with an affinity constant of 0.287 μM as determined in BLI assay (ForteBio Octet Red96e) (QC tested).; Loaded Herceptin on Protein A Biosensor, can bind Cynomolgus CD16, His Tag (BLI verified) (Cat. No. FC6-C52H9) with an affinity constant of 0.31 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Herceptin on FAB2G Biosensor, can bind Cynomolgus CD16, His Tag (BLI verified) (Cat. No. FC6-C52H9) with an affinity constant of 0.23 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q8SPW2-1
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    FCGR3
  • Protein/Peptide Name:
    Fc gamma RIII / CD16
  • Purity:
    >95%
  • Molecular Weight:
    23.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications