Cynomolgus Recombinant CTLA-4 (from HEK293)
Catalog # 103739-368
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Size:25 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human B7-1, Fc Tag (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avitag,His Tag (Cat. No. CT4-C82E5) with a linear range of 2-31 ng/mL (QC tested).; Measured by its binding ability in a functional ELISA. Immobilized Human B7-2, Fc Tag (Cat. No. CD6-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Cynomolgus / Rhesus macaque CTLA-4, Avitag,His Tag (Cat. No. CT4-C82E5) with a linear range of 2-63 ng/mL (Routinely tested).
- Gene ID:G7PL88-1
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:CTLA4|CD152
- Protein/Peptide Name:CTLA-4
- Purity:>92%
- Molecular Weight:15.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103739-368