Order Entry
Puerto Rico
ContactUsLinkComponent
Cynomolgus Recombinant CRTAM (from HEK293)
Cynomolgus Recombinant CRTAM (from HEK293)
Supplier:  ACRO Biosystems
Certificates
Cynomolgus Recombinant CRTAM (from HEK293)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.; Measured by its binding ability in a functional ELISA. Immobilized Human CADM1, His Tag (Cat. No. CA1-H5225) at 5 μg/mL (100 μL/well) can bind Cynomolgus CRTAM, Fc Tag (Cat. No. CRM-C5255) with a linear range of 0.6-10 ng/mL (QC tested).; Loaded Cynomolgus CRTAM, Fc Tag (Cat. No. CRM-C5255) on Protein A Biosensor, can bind Human CADM1, His Tag (Cat. No. CA1-H5225) with an affinity constant of 37.7 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    G7PPC7
  • Carrier-Free:
    Y
  • Protease-free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CD355|CRTAM
  • Protein/Peptide Name:
    CRTAM
  • Purity:
    >90%
  • Molecular Weight:
    56.6 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Nuclease-free:
    Y
  • Shipping Temperature:
    Powder,RT

Specifications

Frequently Bought Together