Recombinant Protein L (from E. coli)
Catalog # 103737-962
Supplier: ACRO Biosystems
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Size:1 g
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in 100 mM NaH2PO4, pH8.5. Normally trehalose is added as protectant before lyophilization.
- Carrier-Free:Y
- Protease-free:Y
- Animal-Free:Y
- Protein Synonyms:RPL|Protein L
- Protein/Peptide Name:Protein L
- Purity:>95%
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 100 mM NaH2PO4, pH8.5. Normally trehalose is added as protectant before lyophilization.
- Nuclease-free:Y
- Shipping Temperature:Powder,RT
- Cat. No.:103737-962