Corning® 3D Clear Penetration Buffer, Corning
Supplier: Corning
Certificates
About this item
Corning 3D Clear Penetration Buffer is used with the Corning 3D immunolabeling process
- Rapid tissue clearing
- Easy to Use
- No Special Equipment Required
- Compatible with IF, FP and other Fluorescent lables
- Microplate and Automation Compatible
- Reversible to Follow up with 2D H&E/IHC
The Corning 3D Clear Penetration Buffer is used with the Corning 3D immunolabeling process and is comprised of PBS / 0.2% Triton™ X-100 / 0.3 M glycine / 20% DMSO
Caution: For best results, refrigerate after opening.
Related Information
Buffer Solutions Recipes for Immunolabeling
Additional Product Details
Corning 3D Clear Tissue Clearing Reagent