HemaPRO Stain Kit, Original; 125 Tests for HP1, Hardy Diagnostics
Catalog # 76422-822
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Description:HemaPRO stain kit
- Cat. No.:76422-822
- No. of reactions:125 test
Specifications
About this item
A Wrights-Giemsa stain, buffer, and rinse for use with the QuickSlide™ HemaPRO automated hematology slide stainer.
- Produces 125 tests per pack in one self-contained package
The pack contains a special stain formulation for enhanced cell quality.