Order Entry
Puerto Rico
ContactUsLinkComponent
QuantiChrom™ Boron Assay Kit, BioAssay Systems
QuantiChrom™ Boron Assay Kit, BioAssay Systems
Catalog # 76423-328
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiChrom™ Boron Assay Kit, BioAssay Systems
Catalog # 76423-328
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DBOR-100
CAS Number:  

Specifications

  • Description:
    Boron assay kit
  • Cat. No.:
    76423-328
  • No. of tests:
    100

Specifications

About this item

Quantitative determination of boron in agricultural and environmental samples.

  • Fast and sensitive: linear detection range: 0.05 to 10 µg/ml (0.05 to 10 ppm) boron with 100 µl sample (96-well)
  • Method: OD 420 nm
  • Samples: water, plant tisse, soil samples, and antibody conjugation solutions

Boron is an essential micronutrient in plants and is involved in maintaining robust cell walls, cell membranes, and reproductive tissues. Although boron is common in the soil in its natural state as a borate mineral, the amount of boron available to plants is actually quite small. As a result, boron deficiency is the second most common micronutrient deficiency among crop plants. In order to keep plant boron levels in a healthy range, supplementation to the soil via fertilizers and additives is often required. If not regulated, a lack of or excess of boron may significantly lower crop yield. In the biotech industry, sodium borohydride is commonly used to conjugate antibodies and typically needs to be removed from the final product, especially for therapeutic antibodies.

BioAssay Systems boron detection kit provides a convenient and reliable means to measure boron. In the assay borate complexes with azomethine-H to create a colored compound that can be measured at 420 nm. This assay can be used with a variety of samples and is simple, sensitive, and adaptable to high-throughput screening.