PrepEngine Sample Preparation Systems, Distek
Supplier: Avantor
Certificates
About this item
Accelerate your content uniformity sample preparation. The PrepEngine was developed through collaborations with leaders in the pharmaceutical and biotech industry to simplify and speed up the content uniformity sample preparation process.
- Ultra-fast sample preparation
- 10 individual stirring stations
- Variety of PrepTube designs and sizes
- Reliable and consistent results
- Adjustable speed and time
The result - a compact benchtop design that incorporates two or ten individually powered, high-output stirring stations which can prepare content uniformity samples up to 90% faster than conventional methods.
Certifications: CE.
Ordering information: Requires PrepTubes (sold separately). Does not include installation or qualification services.
Delivery information: Includes one year comprehensive warranty.