Anti-NP-CoV Rabbit Monoclonal Antibody [clone: 040]
Supplier: Sino Biological
Certificates
About this item
This product is a recombinant monoclonal antibody expressed from HEK293 cells.
"Type:
Antigen:
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: "
Specifications
- Catalog No:
- 103824-926
- Antigen Symbol:
- NP-CoV
- Antigen Name:
- Coronavirus Nucleocapsid
- Antigen Synonyms:
- Nucleoprotein|coronavirus Nucleocapsid|cov np|ncov NP|coronavirus NP|Nucleocapsid|NP|coronavirus Nucleoprotein|novel coronavirus Nucleoprotein
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- 040
- Reactivity:
- SARS-CoV
- Host:
- Rabbit
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- ImmunoFluorescence:
- Yes
- Form:
- Liquid
- Storage Buffer:
- 0.2 μm Filtered solution in PBS
- Storage Temperature:
- This antibody can be stored at 2...8 °C for one month without detectable loss of activity. Antibody products are stable for twelve months from date of receipt when stored at –20...–80 °C. Preservative-Free. Avoid repeated freeze-thaw cycles.
- Shipping Temperature:
- Ambient
- Tested Applications:
- 8