Order Entry
Puerto Rico
ContactUsLinkComponent
Aprotinin
Aprotinin
Supplier:  RPI
Certificates
undefined
Aprotinin
Supplier:  RPI

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    9087-70-1
  • Storage Conditions:
    2...8 °C
  • Enzyme Name:
    Aprotinin
  • Enzyme Activity:
    10000 KIU/ml
  • pH:
    5.0 - 6.4
  • Molecular Formula:
    C₂₈₄H₄₃₂N₈₄O₇₉S₇

Specifications