ContactUsLinkComponent
Overlaminated Identification Tape, 'ESD Caution', UltraTape
Overlaminated Identification Tape, 'ESD Caution', UltraTape
Catalog # 89203-782
undefined
Overlaminated Identification Tape, 'ESD Caution', UltraTape
Catalog # 89203-782
Supplier Number:  0440PT200C3S
Specifications
  • Width:
    2"
  • Color:
    Yellow with black print
  • Length:
    36 yards
  • Material:
    Classic clean
  • Cat. No.:
    89203-782
  • Supplier no.:
    0440PT200C3S
  • Adhesive material:
    Acrylic
Specifications
About this item
Particle-free, residue-free tapes.
UltraTape 'ESD Caution' printed tape provides excellent floor marking, wall marking, equipment marking, and labeling. The printed PVC film is overlaminated to withstand heavy floor traffic and increase chemical resistance.