ContactUsLinkComponent
Polyethylene Low Adhesion Cleanroom Tape, UltraTape
Polyethylene Low Adhesion Cleanroom Tape, UltraTape
Catalog # 89203-788
undefined
Polyethylene Low Adhesion Cleanroom Tape, UltraTape
Catalog # 89203-788
Supplier Number:  1114WH100P3D
Specifications
  • Width:
    1"
  • Color:
    White
  • Length:
    36 yards
  • Material:
    UltraClean
  • Cat. No.:
    89203-788
  • Supplier no.:
    1114WH100P3D
  • Adhesive material:
    Acrylic
Specifications
About this item
Particle-free, residue-free tapes.
UltraTape 1114/0114 is a low adhesion, removable, highly conformable tape designed for cleanroom packaging and in-process work. Its high cleanliness, low outgassing, lack of residue, and excellent ability to conform to surfaces make it ideal for a wide range of cleanroom applications.