ContactUsLinkComponent
Easy Bleach™ Disinfectant Tablets, Berkshire
Easy Bleach™ Disinfectant Tablets, Berkshire
Catalog # 76422-848
Supplier:  Berkshire
undefined
Easy Bleach™ Disinfectant Tablets, Berkshire
Catalog # 76422-848
Supplier:  Berkshire
Supplier Number:  EZBDT101
Specifications
  • Description:
    EZ Bleach™ tablets
  • Size:
    1 tub of approx. 900 tablets (5g)
  • Cat. No.:
    76422-848
Specifications
About this item
NaDCC - Each rapidly dissolving 48.21% sodium dichloro-s-triazinetrione tablet is the equivalent of 29% available chlorine, a broad spectrum and sporicidal disinfectant.
  • No harsh fumes for the end user
  • Less corrosive to surfaces compared to liquid bleach
  • Neutral and consistent pH levels
  • Made in the USA and EPA registered
  • Kills C. difficile spores in 10 minutes, 99.9% effective against respiratory syncytial viruses and influenza H1N1 (colds and flu)
  • Flexibility – Easily control the level of dilution to match your specific disinfectant requirements
1 tub of approx. 900 tablets will produce 190 gallons of liquid bleach.
Tablets have a 3 year shelf life