Order Entry
United States
ContactUsLinkComponent
 
Vivaclear Centrifuge Filters, Sartorius
  76408-740
 :  Sartorius
undefined
Vivaclear Centrifuge Filters, Sartorius
  76408-740
 :  Sartorius
 :  VK01P042

 

  • Pore size:
    0.80 µm
  • Description:
    Vivaclear mini centrifugal filter
  • Material:
    Polyethersulfone/Polypropylene
  • Environmentally Preferable:
  • Max. RCF:
    2000 ×g
  • Cat. No.:
    76408-740

 

 

Vivaclear centrifugal filters are disposable microfiltration devices for the fast and reliable clarification/filtration of biological samples in the range 100 to 500 µl. They can be used in fixed angle rotors suitable for 2.2 ml centrifuge tubes.

  • High-flux polyethersulfone membrane
  • Low hold-up volume (<5 µl) with total length of 43 mm
  • Fast and reproducible performance
  • Rotor: 40 to 45° fixed-angle rotor, 500 µl
  • Filtrate collection tube Ø: 11 mm
  • Body and filtrate collection tube are made up of polypropylene

Applications: Clarification of samples before loading onto Vivapure protein purification spin columns; removal of particles and participates; filtration of plasma and serum; filtration of cells or cell debris.