Order Entry
United States
ContactUsLinkComponent
Accessories for Vivapure® AdenoPACK Kits, Sartorius
Accessories for Vivapure® AdenoPACK Kits, Sartorius
Catalog # 76409-106
Supplier:  Sartorius
CAS Number:  
Accessories for Vivapure® AdenoPACK Kits, Sartorius
Catalog # 76409-106
Supplier:  Sartorius
Supplier Number:  VS-AVPA001
CAS Number:  

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Description:
    Pump tubing set for Vivapure® AdenoPACK™ 100
  • Environmentally Preferable:
  • Cat. No.:
    76409-106
  • Includes:
    Two AdenoPACK 100 syringe filter units with Sartobind ion exchange membrane and all buffers

Specifications

About this item