PIG® White Meltblown Roll, NEW PIG
Catalog # 89208-796
Supplier: New Pig
Specifications
- For Use With:Absorbs oils, fuels, other oil-based liquids only
- Color:White
- Description:Meltblown roll
- Environmentally Preferable:
- Cat. No.:89208-796
- W×L:30" × 150'
- Absorbency:50.7 gal./package
- Supplier no.:MAT863
Specifications
About this item
Economical MeltBlown mat for effective oil response.
- Great performance on water due to fast-wicking fibers
- Highly absorbent MeltBlown polypropylene mat repels water and soaks up only oil and oil-based fluids
- Floats even when completely saturated most effective for marine spills
- Bright white color makes it easy to locate in spill emergencies
- Won't fall apart even when fully saturated
These rolls are ideal for covering large areas.