ContactUsLinkComponent
PIG® Automotive Fluids Cleanup Kit, NEW PIG
  76326-538
 :  New Pig
undefined
PIG® Automotive Fluids Cleanup Kit, NEW PIG
  76326-538
 :  New Pig
 :  KIT5010
  • For Use With:
    Absorbs oils, coolants, solvents, water
  • Description:
    PIG® Automotive fluids cleanup kit
  • Environmentally Preferable:
  • Cat. No.:
    76326-538
Keep your workstations ready for spills with our grab-and-go clean up kit for oils, coolants, paints and more.
  • Compact design lets you store a spill kit virtually anywhere you need one for fast response
  • Easy-open bag protects kit contents from dirt and moisture
  • Includes absorbent mat pads to clean up spilled oils, coolants, solvents, paints and more (absorbs up to 36 ounces)
  • Disposable gloves included to aid cleanup
  • Resealable bag can be used to contain used absorbents
The single-use kit is faster and more efficient than dragging a large spill kit to a small cleanup; safer and more effective than using rags or paper towels.
Only PIG® Spill kits feature PIG® Absorbents.