ContactUsLinkComponent
FDA Approved Digital Oral Thermometer, Propper
  76419-818
 :  Propper
undefined
FDA Approved Digital Oral Thermometer, Propper
  76419-818
 :  Propper
 :  26915500
  • W×D×H:
    19×8×130 mm
  • Response Time:
    60 s
  • Cat. No.:
    76419-818
  • Accuracy:
    ±0.1 °C (35.5...42.0 °C)
  • Range:
    32...42 °C (89.6...107.6 °F)
BT306 digital electronic thermometer is equipped with LCD display with clear reading.
  • Fast and accurate results
  • Easy-to-read temperature in about 60 seconds
  • Last reading memory
  • Fever alarm
  • Auto-shutoff
The accuracy of this thermometer is ±0.2 ºF from 95.9 ºF to 107.6 ºF.