Social Distance Quik Sign Fold-Ups®; Attention, Stop, Accuform®
76417-058
- Description:Social Distance Quik Sign Fold-Ups® floor sign kit
- Dimensions:18×24"
- Cat. No.:76417-058
- Legend:ATTENTION STOP FOR THE SAFETY AND HEALTH
Floor signs alert and remind those to practice social and physical distancing to prevent the spread of COVID-19, the Coronavirus.
- Molded polyethylene plastic stand is 30½H×26"W with feet 22" apart
- Includes one 18H×24"W corrugated plastic printed sign that fits into slot
- Handle holes for easy portability
Stand closes to 2" thick with molded-in lug points for nesting into each other when stacked.