Bertin Instruments White Blocking Plate, 2 ml, Bertin Corp
76415-488
- Description:White blocking plate, 2 ml
- Cat. No.:76415-488
- For use with:Precellys 24
White blocking plate for Precellys 24.
- Required to process 2 ml tubes
Allows you to process from 1 to 24, 2 ml samples with the Precellys Evolution.