Orders LinkContactUsLinkComponent
Bertin Instruments User Maintenance Kit for Precellys Evolution, Bertin Corp
Bertin Instruments User Maintenance Kit for Precellys Evolution, Bertin Corp
Precellys Evolution maintenance kit
Catalog # 76415-486
Supplier:   BERTIN CORP
Supplier #:  S000808-PEVO0-A.0
Bertin Instruments User Maintenance Kit for Precellys Evolution, Bertin Corp
Catalog # 76415-486
Supplier:  BERTIN CORP
Bertin Instruments User Maintenance Kit for Precellys Evolution, Bertin Corp
Precellys Evolution maintenance kit
Price
$659.82
Log in for availability
Kit of 1
Minimum order quantity is 1
Product Details & Documents
Precellys Evolution maintenance kit contains all the external pieces to replace each year of use allowing to give the maximum life time to the equipment.
  • Allows you to enhance the life time of your Precellys Evolution equipment
For Precellys Evolution, Contains: 2× vacuum joints, 1× containment seal, 1× anti-rotation tubing kit.
Specifications
  • Description:
    Precellys Evolution maintenance kit