Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cryolys Evolution, Bertin Corp

Cryolys Evolution, Bertin Corp

Cryolys® Evolution, 335W×335L×100H mm, 42.6 kg (5.7 lbs.) (full unit); 1.3 kg (2.8 lbs.) (cooling module)
Catalog # 76415-478
Supplier:   BERTIN CORP
Supplier #:  P000671-CLYS2-A.0
Cryolys Evolution, Bertin Corp
Catalog # 76415-478
Supplier:  BERTIN CORP
Cryolys Evolution, Bertin Corp
Cryolys Evolution, Bertin Corp

Cryolys® Evolution, 335W×335L×100H mm, 42.6 kg (5.7 lbs.) (full unit); 1.3 kg (2.8 lbs.) (cooling module)

Price
$9,318.76
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

The Cryolys Evolution is the cooling system specially developped for Precellys Evolution tissue homogenizer.

  • Easy to install: Powered and controlled by Precellys Evolution, no compressed air required
  • Easy to use: Precellys Evolution software automatically monitors the protocol run
  • Fast and accurate:Target temperature reached in less than two minutes and fully monitored
  • Pressure: 5 to 116 psi
  • Includes one-year warranty

Cryolys Evolution is the ideal solution when working on native state thermo-sensitive molecules such as active RNA, siRNA, metabolites, enzymes, proteins while getting an hight yield extraction.

It allows an optimal protection of the sensitive molecules from heat degradation. Based on dry ice sublimation, it enhances the efficiency of molecular extraction, resulting in premium quality analysis, maintaining the desired temperature between 0 and 10 °C before and during the tissue homogenization.

Space requirement: Height over bench: 70 cm; Depth of the bench: 70 cm

Certifications: CE certified.

Ordering information: Unit plugged to a Precellys Evolution is required.

Specifications
  • Description:
    Cryolys® Evolution
  • Dimensions:
    335W×335L×100H mm
  • Weight:
    42.6 kg (5.7 lbs.) (full unit); 1.3 kg (2.8 lbs.) (cooling module)
  • Cat. No.:
    76415-478
  • Capacity:
    1.5 kg (3.3 lbs.) of dry ice
  • Run Time:
    1.5 kg (3.3 lbs.) of dry ice for 30 min
  • Sound Level:
    <60 dB
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.