Biotin-20-UTP 10 mM in buffer (Tris-HCl, pH 7.5)
Catalog # 89139-100
Supplier: Biotium
CAS Number:
Specifications
- Pack type:Plastic vial
- Size:25 µl
- Cat. No.:89139-100
- Supplier no.:40034
Specifications
About this item
:
:1036.26 g/mol
Storage Temperature:Freezer
Formula:C₃₃H₅₁N₆O₂₂P₃SLi₄