Biotin-11-CTP (Biotin-11-cytidine-5′-triphosphate, tetralithium salt) 10 mM in buffer (TRIS-HCl pH 7.5)
Catalog # 89139-102
Supplier: Biotium
CAS Number:
Specifications
- Pack type:Plastic vial
- Size:25 µl
- Cat. No.:89139-102
- Supplier no.:40035
Specifications
About this item
:
:901.42 g/mol
Formula:C₂₈H₄₂N₇O₁₇P₃SLi₄