Specifications
- Description:BCR® Large mop head frame with plastic eyes
- Dimensions:58×9 cm (23×3.5")
- Material:Aluminum, anodized
- Cat. No.:76417-138
Specifications
About this item
This flat mop head is light and easy to maneuver, even with wet wipers attached. The frame is equipped with plastic eyes for easy and efficient wiper attachment.
- Clean, light, and durable
- High-quality anodized aluminum
- Autoclavable
- Plastic eyes for wiper attachment
Efficient cleaning of floors, walls, and other large surfaces is critical to the overall cleanliness of the production environment. BCR® Mop systems replicate the most effective way to clean a surface-wiping. Berkshire’s mop systems are easy to use, highly effective, and quick. The system is very flexible, combining a wide range of alternative mop wipers designed for particular levels of cleanliness with a range of flat mop heads.
The plastic eyes hold multiple mop wipers in place and are very easy to operate. The bottom of the mop is smooth and has no lines or grooves where bacteria or other contamination can settle. Telescopic handles are available separately.