Order Entry
Puerto Rico
ContactUsLinkComponent
Cell Freezing Medium 50 ml, Cell Systems
Cell Freezing Medium 50 ml, Cell Systems
Catalog # 103810-702
Supplier:  Cell Systems
CAS Number:  
undefined
Cell Freezing Medium 50 ml, Cell Systems
Catalog # 103810-702
Supplier:  Cell Systems
Supplier Number:  4Z0705
CAS Number:  

Specifications

  • Description:
    Reagent
  • Size:
    50 ml
  • Cat. No.:
    103810-702

Specifications

About this item

Optimize the plating, passaging and freezing of cell cultures with Cell Systems Reagents. Growth supplements and antibiotic products are also available to ensure each experiment is initiated in an optimal environment.

  • High quality specialized reagents
  • Always available
  • Overnight delivery

Cell Systems Cell Freezing Medium is a specialized medium providing the optimal environment for the freeze/thaw cycle of cell cultures and assists in minimizing cellular damage during the process. Cell Systems Cell Freezing Medium (1X) 50 ml. Contains Conditioned Cell Systems Media.

Use of the Passage Reagent Group™ (PRG) to release cells from adherent culture for freezing is critically important in minimizing membrane and cytoskeletal damage. Cell membranes are materially and cumulatively damaged by detachment or when subjected to the stress of freezing and thawing. Use of PRG in conjunction with Cell Freezing Medium and Attachment Factor™ minimizes cellular stress and damage associated with these processes and results in improved viability post-thaw.

Handling and Storage:
Store refrigerated at 2 to 8°C. Once opened, refrigerated shelf life is 30 days.

Certifications: Cell Systems media and reagents are Sterile, made with WFI and all components are cGMP and ISO Compliant.