ContactUsLinkComponent
THE SYSTEM™, Cell Systems
THE SYSTEM™, Cell Systems
Catalog # 103810-686
Supplier:  Cell Systems
CAS Number:  
undefined
THE SYSTEM™, Cell Systems
Catalog # 103810-686
Supplier:  Cell Systems
Supplier Number:  CSSA101
CAS Number:  
Specifications
  • Description:
    Cell culture media
  • Size:
    Kit
  • Cat. No.:
    103810-686
Specifications
About this item
THE SYSTEM™ is an all-in-one cell growth and passage system designed to provide your lab with convenience and consistency.
  • Save money with an All-in-One Kit
  • Optimize your experiments
  • Gain consistency in your research
  • High quality specialized media and reagents
  • Always available
  • Overnight delivery
  • Always available
  • Overnight delivery
The complete cell culture kit to optimize your experiments. The System™ includes: 2×500 ml Cell Systems Classic Complete Media (4Z0-500), 1×100 ml Attachment Factor™, 2×5 ml CultureBoost™, one set of 100 ml Passage Reagent Group™ and 1×50 ml Cell Freezing Medium™.
Medium Component:
Medium - formulated with 10% serum. This medium becomes complete once activated with the included CultureBoost™ supplement. 4Z0-500 is certified and intended for experimental application.
Growth Supplement Component:
CultureBoost™ is the broad-spectrum supplement used to activate Cell Systems Complete Medium. CultureBoost™ contains Cell Systems bovine Growth Factor and porcine heparin.
PRG Component:
Passage Reagent Group™ (PRG) is a matched set of Cell Systems Certified™ reagents for releasing cells from culture for subculture or freezing. The PRG contains three parts: PRG-1(EDTA-dPBS Solution), PRG-2 (Trypsin/EDTA-dPBS Solution) and PRG-3 (Trypsin Inhibitor-dPBS Solution). The chelating agent EDTA in PRG-1 prepares for PRG-2, which contains highly purified trypsin. PRG-3 inactivates the protease in PRG-2 and stabilizes the cell membranes.
Cell Freezing Medium™ Component:
Cell Systems Cell Freezing Medium™ is a specialized media, which when used in conjunction with Passage Reagent Group™ provides a beneficial environment for the freeze/thaw cycle of cell cultures, assisting in the minimization of cellular damage during the process.
Attachment Factor™ Component:
Attachment Factor™ is an extracellular matrix (ECM) product that promotes cell attachment to tissue culture surfaces and encourages correct polarity and cytoskeletal organization. Use of Attachment Factor™ is critically important when cultures are to be initiated, grown, passaged, or used within the Cell Systems Medium family.
Certifications: Cell Systems media and reagents are Sterile, made with WFI and all components are cGMP and ISO Compliant.