Order Entry
Puerto Rico
ContactUsLinkComponent
Medium Kit With Serum and Without Growth Factor, Cell Systems
Medium Kit With Serum and Without Growth Factor, Cell Systems
Catalog # 103810-618
Supplier:  Cell Systems
CAS Number:  
undefined
Medium Kit With Serum and Without Growth Factor, Cell Systems
Catalog # 103810-618
Supplier:  Cell Systems
Supplier Number:  4Z0500S
CAS Number:  

Specifications

  • Description:
    Cell culture media
  • Size:
    500 ml
  • Cat. No.:
    103810-618

Specifications

About this item

Our suite of specialized media provides our customers with a diverse set of tools to enhance research through consistent and stable environments for cell cultures. Cell Systems media may be used with all primary, immortalized, and established cell lines.

  • Based on a modified and optimized basal DMEM/F12 (1:1)
  • Contain serum at 10% by volume
  • Do not contain phenol red
  • Packaged in 500mL bottles (as a kit)

Medium is formulated with 10% serum. Kit consists of 500mL 4Z0-500-S Cell Systems Medium (formulated without growth factor) and 10mL Attachment Factor™ (extracellular matrix reagent for coating flasks). This is not a complete medium. For complete medium, add animal derived growth factor (CultureBoost™, 4CB-500) or human recombinant growth factor (CultureBoost-R™ 4CB-500-R). 4Z0-500-S is Certified and intended for experimental application.

Cell Systems Medium contains no added hormones.

Cell Systems Medium contains no antibiotics; however, Bac-Off® (Cat. #4Z0-643), is the recommended antibiotic for all Cell Systems medium. Bac-Off® is at or above the [2ng/mL] MIC of aerobic gram-positive and gram-negative microorganisms.

Cell Systems Medium contains no phenol red. Phenol red in cell culture medium binds to receptors on cell surfaces and causes the cell to respond as though pharmacologic levels of estrogen and/or inflammatory mediators (such as prostaglandin or thromboxane A2) are present in the medium. We exclude this unnecessary additive in order to optimize experimental conditions for researchers.

Certified for use with more than 100 human and animal cell cultures and cell lines including:

Human: All Cell Systems Primary Cells; Bone Marrow Cells (CFU-F and Erythroid Burst Cultures); Connective Tissue Cells; Dermal Fibroblast Cells; Endothelial Cells (Aortic, Arterial, Coronary Artery, Vascular and Venous); Fetal Lung Cells; Embryonic and Adult Stem Cells; Fibroblast Cells; Glomerular Mesangial Cells; Immortalized / Tumor-derived Mesenchymal Cells; Microvascular Cells (Cerebral, Coronary, Dermal, Glomerular, Liver, Lung and Retinal); Smooth Muscle Cells; Tumor Neovascular Cells

Storage: If the entire unit will not be used within 30 days, aliquot and freeze in smaller units which will be used within 30 days. Otherwise, immediately refrigerate the medium. Once opened, the shelf life is 30 days at refrigerated (2-8°C) temperatures.

For best results, use Cell Systems Medium in conjunction with cell culture vessels coated with Attachment Factor™ extracellular matrix reagent.

Certifications: Cell Systems media and reagents are Sterile, made with WFI and all components are cGMP and ISO Compliant.