Order Entry
Puerto Rico
ContactUsLinkComponent
Cell Systems Human Liver Sinusoidal Endothelial Cells, Primary Cell and Media Company
Cell Systems Human Liver Sinusoidal Endothelial Cells, Primary Cell and Media Company
Catalog # 103810-664
Supplier:  Cell Systems
CAS Number:  
undefined
Cell Systems Human Liver Sinusoidal Endothelial Cells, Primary Cell and Media Company
Catalog # 103810-664
Supplier:  Cell Systems
Supplier Number:  ACBRI566
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Single Donor/Pooled:
    Single donor
  • Description:
    Human liver sinusoidal endothelial cells
  • Size:
    Vial
  • Storage Conditions:
    Upon receipt, store in Liquid Nitrogen
  • Shipping Conditions:
    Shipped on Dry Ice
  • Storage Buffer:
    Frozen in Cell Systems Cell Freezing Medium (4Z0705)
  • Cat. No.:
    103810-664

Specifications

About this item

Our primary human cells offer unmatched consistency and a more biologically relevant tool than other cell types. Our cells are antibody-free and come from a deep inventory to help researchers with reproducibility and improved human insights.

  • Highly proliferative and healthy (~85% viability)
  • Antibody free and more biologically relevant
  • Offered with high cell count (~1 million cells per vial)
  • Well-cited in scientific journals
  • Ideal for 2D and 3D cultures, microfluidic devices and co-cultures

Primary Human Liver Sinusoidal Microvascular Endothelial Cells (ACBRI566) were initiated from normal human liver tissue by elutriation following dispase treatment of tissue. This cell is antibody free. Each vial will initiate a 25 to 75 cm² cell culture. These primary cells are isolated, frozen and qualified using Cell Systems Certified Media and Reagents--which are specified for use in the research environment. Cell Systems primary cells are exclusively from normal, site-specific human tissue specimen, and available in frozen vial or proliferating flask.

These cells were originated using Cell Systems Complete Serum-Free Medium (SF-4Z0-500), and subsequently grown and passaged in Cell Systems Complete Medium (4Z0-500). They are available at Passage 3 (< 12 cumulative population doubling)] cryopreserved in Cell Systems Cell Freezing Medium™ (4Z0-705). This vial will initiate a Passage 4 cell culture in a 75 cm² flask. Each vial contains more than 1×106 cells.

Each vial of cells is shipped with vials of Bac-Off® (antibiotic) and CultureBoost™ (animal derived growth factors) or CultureBoost-R™ (human recombinant growth factors) at no additional cost.

Standard Tests:
HIV Serologic Test (donor level HIV AB EIA) - Negative
HIV PCR Test (frozen cell pool by CLIA Licensed Clinical Lab) - Negative
Mycoplasma spp. (frozen cells) (ATCC method by CLIA Licensed Clinical Lab) - Negative.

Characterization:
Inducible expression of CD 62E (E-Selectin) >90% positive by immunofluorescence
Cytoplasmic VWF / Facor VIII > 95% positive by immunofluorescence
Cytoplasmic upake of Ki-l-AC-LDL > 95% positive by immunofluorescence.

Caution: For Research Use Only