- Cell Format:Cryopreserved
- Single Donor/Pooled:Pooled
- Description:Human pancreatic microvascular endothelial cells
- Size:Vial
- Storage Conditions:Upon receipt, store in Liquid Nitrogen
- Shipping Conditions:Shipped on Dry Ice
- Storage Buffer:Frozen in Cell Systems Cell Freezing Medium (4Z0705)
- Cat. No.:103810-640
Our primary human cells offer unmatched consistency and a more biologically relevant tool than other cell types. Our cells are antibody-free and come from a deep inventory to help researchers with reproducibility and improved human insights.
- Highly proliferative and healthy (~85% viability)
- Antibody free and more biologically relevant
- Offered with high cell count (~1 million cells per vial)
- Well-cited in scientific journals
- Ideal for 2D and 3D cultures, microfluidic devices and co-cultures
Primary Human Pancreatic Microvascular Endothelial Cells (ACBRI134) were initiated by elutriation from dissociated normal human pancreatic tissue. This cell is antibody free. Each vial will initiate a 25 to 75 cm² cell culture. These primary cells are isolated, frozen and qualified using Cell Systems Certified Media and Reagents--which are specified for use in the research environment. Cell Systems primary cells are exclusively from normal, site-specific human tissue specimen, and available in frozen vial or proliferating flask.
These cells were originated using Cell Systems Complete Serum-Free Medium (SF-4Z0-500), and subsequently grown and passaged in Cell Systems Complete Medium (4Z0-500). They are available at Passage 3 (< 12 cumulative population doublings) cryopreserved in Cell Systems Cell Freezing Medium™ (4Z0-705). This vial will initiate a Passage 4 cell culture in a 75 cm² flask. Each vial contains more than 1×106 cells.
Each vial of cells is shipped with vials of Bac-Off® (antibiotic) and CultureBoost™ (animal derived growth factors) or CultureBoost-R™ (human recombinant growth factors) at no additional cost.
Standard Tests:
HIV Serologic Test (donor level HIV AB EIA) -Negative
HIV PCR Test (frozen cell pool by CLIA Licensed Clinical Lab) - Negative
Mycoplasma spp. (frozen cells) (ATCC method by CLIA Licensed Clinical Lab) - Negative.
Characterization:
Expression of CD 31 > 98% positive by immunofluorescence
Cytoplasmic Vimentin Intermediate Filaments > 98% positive by immunofluorescence
Cytoplasmic VWF / Factor VIII > 95% positive by immunofluorescence
Fibroblast Surface Protein < 2% positive by immunofluorescence.