Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Silver nitrate 20% in aqueous solution

Silver nitrate 20% in aqueous solution

500 ml
Catalog # 101410-792
Supplier #:  21054-20
Silver nitrate 20% in aqueous solution
Catalog # 101410-792
CAS Number:  7761-88-8
Silver nitrate 20% in aqueous solution
Silver nitrate 20% in aqueous solution

500 ml

CAS Number:  7761-88-8
Price
$642.89
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Silver Nitrate solution is available in concentrations of 1.0% aqueous, 2.0% aqueous, 5.0% aqueous, 10.0% aqueous, and 20.0% aqueous.

Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • UN:1493
  • ADR:5.1,II
Specifications

Specifications

  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.