HiScreen™ Capto PlasmidSelect, Columns, Cytiva
Catalog # 76325-064
Supplier: Cytiva
Specifications
- Flow rate:0.8 - 2.3 ml/min
- Max. Pressure:8 bar
- Capacity:4.7 ml
- Description:HiScreen™ capto plasmid select
- Bed dimensions:7.7×100 mm
- Cat. No.:76325-064
Specifications
About this item
Columns packed with Capto PlasmidSelect are suitable for purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications. Capto PlasmidSelect is a chromatography resin that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
- High-flow purification of supercoiled DNA from research scale to cGMP production
- Flexible process design due to a large operational window of flow velocities and bed heights
- Higher throughput for improved productivity and process economy
- Hydrophilic properties of base matrix prevent non-specific binding
- BioProcess resin supported for industrial applications
- Convenient small-scale purification, process development, and scale-up using prepacked HiScreen™ columns
Capto PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production.