Purified, concentrated, high quality silver nps; sterile and endotoxin-free. Bare citrate surface.
- High quality nanoparticles, precisely engineered and highly characterized
- Unagglomerated and monodisperse
- High concentration materials (1 mg/ml, 9.27 mm, 1000 ppm, ~40 to 135 OD [depending on size])
- Extensively purified to remove residual reactants
- Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
- BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
- "Bare" citrate surface is stable and displaceable
Spherical silver nanoparticles, 1 mg/ml (high concentration). Citrate offers stable and readily displaceable surface. BioPure materials are Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <15% for 30 to 200 nm sizes, <20% for 20 nm, and <25% for 5 and 10 nm sizes).
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.