High quality silver nanoparticles. Provided with full COA; great value. Displaceable citrate surface
- High quality nanoparticles, precisely engineered and highly characterized
- Unagglomerated and monodisperse
- Standard colloidal concentration (0.02 mg/ml, 20 ppm, 0.185 mm, OD's of 0.8 to 2.7 [depending on size])
- Citrate surface is stable and displaceable
Spherical silver nanoparticles, 0.02 mg/ml, Citrate surface is stable and displaceable. NanoXact materials are precisely engineered, unagglomerated nanospheres with a narrow size distributions (CVs <15% for 30 to 200 nm sizes, <20% for 20 nm, and <25% for 5 and 10 nm sizes).
Each batch is extensively characterized to ensure it meets the highest quality standards so you can achieve reliable and reproducible results. Products are supplied with COA's that include batch-specific TEM, DLS, Zeta Potential, UV-Vis, and pH measurements.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.