TransPorter® Transport Swab, Cary Blair
Catalog # 76377-444
Supplier: Hardy Diagnostics
Specifications
- Format:Rayon tip swab
- Description:Cary blair, single swab, plastic shaft
- Cat. No.:76377-444
- Packaging:50 per box
- Cap color:Red
Specifications
About this item
Cary-Blair medium is a modification of the original stuart formulation, but is almost exclusively used for the transportation of rectal swabs and fecal specimens for the isolation of enteric pathogens.
- COPAN carries Cary-Blair medium agar gel