Rat Recombinant IL-3 B (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Interleukin 3 beta (IL-3 β) is a cytokine that is produced by activated T cells and mast cells. IL-3 β induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 β also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
- High quality
- Low endotoxin
- Affordable
Caution: For Research use only and is not for use in humans.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Rat
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1.0×10⁴ units/mg
- Gene ID:P04823
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:MCGF|P-cell stimulating factor|HCGF|Multi-CSF
- Protein/Peptide Name:IL-3 B (AF)
- Purity:≥95%
- Molecular Weight:16.3 kDa
- Sequence:MISDRGSDAH HLLRTLDCRT IALEILVKLP YPQVSGLNNS DDKANLRNST LRRVNLDEFL KSQEEFDSQD TTDIKSKLQK LKCCIPAAAS DSVLPGVYNK DLDDFKKKLR FYVIHLKDLQ PVSVSRPPQP TSSSDNFRPM TVEC
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Frequently Bought Together