Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Amphiregulin (Animal Free) (from E. coli)
Human Recombinant Amphiregulin (Animal Free) (from E. coli)
New Product
Certificates
undefined
Human Recombinant Amphiregulin (Animal Free) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: 3T3 Cell Proliferation , ED50 ≤20 ng/ml, Bioactivity ≥5.0×10⁴ units/mg
  • Gene ID:
    P15514
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    AR|Colorectum cell-derived growth factor|CRDGF|AREG
  • Protein/Peptide Name:
    Amphiregulin
  • Purity:
    ≥95%
  • Molecular Weight:
    10.1 kDa
  • Sequence:
    SVRVEQVVKP PQNKTESENT SDKPKRKKKG GKNGKNRRNR KKKNPCNAEF QNFCIHGECK YIEHLEAVTC KCQQEYFGER CGEKSMK
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Related Information