Pig Recombinant FLT-3 Ligand (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation. FLT-3 ligand signaling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor. FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3).
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Pig
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: OCI-Aml5 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2.0×10⁵ units/mg
- Gene ID:D2K7D6
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Flt3L|Fms-related tyrosine kinase 3 ligand
- Protein/Peptide Name:FLT-3 Ligand
- Purity:≥95%
- Molecular Weight:17.3 kDa
- Sequence:MSPDCSFPHS PISSTFANTI RQLSDYLLQD YPVTVASNLQ DDELCGAFWR LVLAQRWMGQ LKTVAGSQMQ KLLEAVNTEI VFVTSCALQP LPSCLRFVQA NISHLLQDTS QQLVALKPWI TRRNFSRCLE LQCQPDPSTL LPPRSPGALE ATSLP
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay