Order Entry
Puerto Rico
ContactUsLinkComponent
Dog Recombinant RC GM-CSF (Animal free) (from E. coli)
Dog Recombinant RC GM-CSF (Animal free) (from E. coli)
Certificates
undefined
Dog Recombinant RC GM-CSF (Animal free) (from E. coli)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Dog
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤15 ng/ml, bioactivity ≥6.7×10⁴ units/mg
  • Gene ID:
    P48749
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    MGI-1GM|CSF-2|Pluripoietin-α
  • Protein/Peptide Name:
    RC GM-CSF (AF)
  • Purity:
    ≥95%
  • Molecular Weight:
    14.4 kDa
  • Sequence:
    MAPTRSPTLV TRPSQHVDAI QEALSLLNNS NDVTAVMNKA VKVVSEVFDP EGPTCLETRL QLYKEGLQGS LTSLKNPLTM MANHYKQHCP PTPESPCATQ NINFKSFKEN LKDFLFNIPF DCWKPVKK
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Related Information